missing translation for 'onlineSavingsMsg'
Learn More

Chlorpropamide

Product Code. 15476209
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15476209 100 g 100g
15466209 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15476209 Supplier Thermo Scientific Alfa Aesar Supplier No. J64110.22
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Increases insulin secretion | CAS: 94-20-2 | C10H13ClN2O3S | 276.735 g/mol

Chlorpropamide is used in the treatment of diabetes metilus type 2. It acts to increase the secretion of insulin and is not effective in patients who do not have pancreatic beta cell function.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorpropamide is used in the treatment of diabetes metilus type 2. It acts to increase the secretion of insulin and is not effective in patients who do not have pancreatic beta cell function.

Solubility
Soluble in ethanol (1:12), acetone (1:5), chloroform (1:9) and solutions of alkali hydroxides. Does not mix well with water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 94-20-2
Molecular Formula C10H13ClN2O3S
Molecular Weight (g/mol) 276.735
MDL Number MFCD00079004
InChI Key RKWGIWYCVPQPMF-UHFFFAOYSA-N
Synonym chlorpropamide, chloropropamide, diabinese, chlorpropamid, diabenese, glucamide, meldian, chlorodiabina, chloronase, diabeneza
PubChem CID 2727
ChEBI CHEBI:3650
IUPAC Name 1-(4-chlorophenyl)sulfonyl-3-propylurea
SMILES CCCNC(=O)NS(=O)(=O)C1=CC=C(C=C1)Cl

Specifications

Color White
Quantity 100 g
Merck Index 14,2186
Solubility Information Soluble in ethanol (1:12),acetone (1:5),chloroform (1:9) and solutions of alkali hydroxides. Does not mix well with water.
Formula Weight 276.74
Physical Form Powder
Chemical Name or Material Chlorpropamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.