missing translation for 'onlineSavingsMsg'
Learn More

Chlorprothixene, MedChemExpress

Product Code. 16494317
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
50mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16494317 50 mg 50mg
16458206 100 mg 100mg
16421406 200 mg 200mg
16412086 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16494317 Supplier Medchem Express Supplier No. HYB0274
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Chlorprothixene is a dopamine and histamine receptors antagonist with Kis of 18 nM, 2.96 nM, 4.56 nM, 9 nM and 3.75 nM for hD1, hD2, hD3, hD5 and hH1 receptors, respectively. Antipsychotic activity.

Chlorprothixene is a dopamine and histamine receptors antagonist with Kis of 18 nM, 2.96 nM, 4.56 nM, 9 nM and 3.75 nM for hD1, hD2, hD3, hD5 and hH1 receptors, respectively. Antipsychotic activity.

Specifications

Chemical Name or Material Chlorprothixene
Physical Form Solid
CAS 113-59-7
Color Earth Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C18H18ClNS
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information DMSO : 33.33 mg/mL (105.52 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES CN(C)CC/C=C1C2=C(SC3=C\1C=CC=C3)C=CC(Cl)=C2
Molecular Weight (g/mol) 315.86
Formula Weight 315.86
Percent Purity 99.13%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.