missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Chlortetracycline hydrochloride, can be used as secondary standard

Product Code. 10115160 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mg
2.5 g
Packaging:
Glass Bottle
Unit Size:
2.5g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10115160 500 mg Glass Bottle 500mg
11916911 2.5 g Glass Bottle 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10115160 Supplier Thermo Scientific Chemicals Supplier No. 268245000
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chlortetracycline hydrochloride, CAS # 64-72-2, is a broad-spectrum antibiotic of the tetracycline family

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Chlortetracycline hydrochloride is a broad-spectrum antibiotic of the tetracycline family
  • Receives its name from a 7-chloro substitution on one of the rings of its parent structure, tetracycline
  • A bacteriostatic that inhibits the 30S ribosomal subunit and prevents peptide formation
  • Can work as an antiprotozoal and antifungal

Applications

  • Chlortetracycline is suitable for use in biochemistry, microbiology, in vivo research, and cell culture
  • Intended for research use only
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64-72-2
Molecular Formula C22H24Cl2N2O8
Molecular Weight (g/mol) 515.34
InChI Key QYAPHLRPFNSDNH-CIVPRPTRSA-N
Synonym CTC
PubChem CID 66577600
IUPAC Name (4S,4aR,5aS,6S,12aR)-7-chloro-4-(dimethylamino)-1,6,10,11,12a-pentahydroxy-6-methyl-3,12-dioxo-4,4a,5,5a-tetrahydrotetracene-2-carboxamide;hydrochloride
SMILES CC1(C2CC3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C(C=CC(=C41)Cl)O)O)O)O)C(=O)N)N(C)C)O.Cl

Specifications

Type Chlortetracycline Hydrochloride
Physical Form Fine Crystalline Powder
Melting Point 220.0°C
Infrared Spectrum Authentic
Packaging Glass Bottle
Specific Rotation − 242.50 (20.00°C c=0.5,water)
Color Yellow
Quantity 500 mg
Solubility Information Solubility in water: 8.6g/L (28°C). Other solubilities: practically insoluble in acetone, ether,, chloroform, dioxane and propylene glycol, 17.4g/L methanol (28°C), 1.7g/L ethanol (28°C), soluble in solutions of alkali hydroxides and, carbonate
Formula Weight 515.33
Percent Purity ≥80 wt% (as is, calculated) (HPLC)
Chemical Name or Material Chlortetracycline hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • Chlortetracycline hydrochloride
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.