missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chlortetracycline hydrochloride, MedChemExpress
Description
Chlortetracycline hydrochloride (7-Chlorotetracycline hydrochloride) is a specific and potent calcium ionophore antibiotic, inhibits binding of aminoacyl-tRNA to ribosomes.
Specifications
Specifications
| Chemical Name or Material | Chlortetracycline hydrochloride |
| Physical Form | Solid |
| CAS | 64-72-2 |
| Color | Yellow |
| Quantity | 250 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C22H24Cl2N2O8 |
| For Use With (Application) | COVID-19-immunoregulation |
| Synonym | 7-Chlorotetracycline hydrochloride |
| Solubility Information | H2O : 6.66 mg/mL (12.92 mM; Need ultrasonic and warming) ∣DMSO : < 1 mg/mL (insoluble or slightly soluble) |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Hittar du en möjlighet till förbättring?