missing translation for 'onlineSavingsMsg'
Learn More

Chlorzoxazone, MedChemExpress

Product Code. 16451118
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16451118 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16451118 Supplier Medchem Express Supplier No. HYB1462
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Chlorzoxazone is a centrally acting muscle relaxant used to treat muscle spasm and the resulting pain or discomfort.

Chlorzoxazone is a centrally acting muscle relaxant used to treat muscle spasm and the resulting pain or discomfort.

Specifications

Chemical Name or Material Chlorzoxazone
CAS 95-25-0
Color White
Purity Grade Notes Research
Molecular Formula C7H4ClNO2
Quantity 5 g
Solubility Information DMSO : ≥ 100 mg/mL (589.73 mM)
SMILES O=C1OC2=CC=C(Cl)C=C2N1
Molecular Weight (g/mol) 169.57
Formula Weight 169.57
Percent Purity 99.6%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.