missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHM Antibody (SR2298), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22495-100ul
This item is not returnable.
View return policy
Description
CHM Monoclonal antibody specifically detects CHM in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence
Specifications
| CHM | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunofluorescence | |
| SR2298 | |
| Western Blot 1:500-1:2000, Immunocytochemistry/Immunofluorescence 1:50-1:200 | |
| choroideraemia protein, choroideremia (Rab escort protein 1), Rab geranylgeranyltransferase component A, rab proteins geranylgeranyltransferase component A 1, REP1 | |
| A synthesized peptide derived from human CHM (Uniprot #: P24386) | |
| 100 μg | |
| Neuroscience, Signal Transduction | |
| 1121 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction