missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CHML Recombinant Mouse Monoclonal Antibody (K92072_16C11)

Product Code. 30828545
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30828545 100 μL
30830359 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30828545 Supplier Invitrogen™ Supplier No. MA564477

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Recombinant Monoclonal Antibody

Immunogen sequence: VVIIGTGLPE SILAAACSRS GQRVLHIDSR SYYGGNWASF SFSGLLSWLK EYQQNNDIGE ESTVVWQDLI HETEEAITLR KKDETIQHTE AFCYASQDME DNVEEIGALQ KNPSLGVSNT FTEVLDSALP EESQLSYFNS DEMPAKHTQK SDTEISLEVT DVEESVEKEK YCGDKTCMHT VSDKDGDKDE SKSTVEDKAD EPIRNRITYS QIVKEGRRFN Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 56%.

Binds unprenylated Rab proteins, presents it to the catalytic component B, and remains bound to it after the geranylgeranyl transfer reaction. The component A may be regenerated by transferring its prenylated Rab to a protein acceptor. Less effective than REP-1 in supporting prenylation of Rab3 family.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CHML
Applications Western Blot
Classification Recombinant Monoclonal
Clone K92072_16C11
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
Gene CHML
Gene Accession No. P26374
Gene Alias AI450814; CHM like, Rab escort protein 2; Chml; choroideraemia-like protein; choroideremia-like; choroideremia-like (Rab escort protein 2); choroideremia-like protein; E030003F13Rik; rab escort protein 2; rab escort protein-2; rab proteins geranylgeranyltransferase component A 2; REP2; REP-2
Gene Symbols CHML
Host Species Mouse
Immunogen Recombinant human CHML (aa 11-230).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1122
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.