Learn More
Berkshire™ Choice™ Cleanroom Face Masks
Product Code. 18849410 Shop All Berkshire Products
Change view
Click to view available options
Packaging Type:
Bulk Packaged
Product Code. Packaging Type
18849410 Bulk Packaged
1 options
Product Code. 18849410 Supplier Berkshire™ Supplier No. CHFM18040B
Three-layer cleanroom face masks provide filtration efficiency while providing long wearing comfort.
  • 3-Layer cleanroom masks
  • Knitted nylon earloops
  • Filter medium: meltblown polypropylene provides particle and bacterial filtration
  • Fully enclosed nosepiece is adjustable
  • Available in bulk or individually wrapped packaging
Specifications
Product Type Cleanroom Face Mask
Color White
Fastening Type Earloops
Material Polypropylene
Sterility Non-sterile
Style Pleated
Certifications/Compliance ASTM F2299
Certifications/Compliance (Europe) EN 14683
Cleanroom Class Class 10 (ISO 4)
Construction 3-ply
Disposable Single-use
Filter Efficiency 99.7% (PFE), 99.7% (BFE)
Model CHFM18040B
Packaging Quantity 50/Pk., 40 Pk./Cs.
Packaging Type Bulk Packaged
Series Choice™