missing translation for 'onlineSavingsMsg'
Learn More

Cholesterol, Thermo Scientific™

Product Code. 15845108 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 g
100 g
Packaging:
Plastic bottle
Unit Size:
100g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15845108 100 g Plastic bottle 100g
15835108 5 g Plastic bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15845108 Supplier Thermo Scientific Supplier No. J13580.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 57-88-5
Molecular Formula C27H46O
Molecular Weight (g/mol) 386.664
MDL Number MFCD00003646
InChI Key HVYWMOMLDIMFJA-DPAQBDIFSA-N
Synonym cholesterol, cholesterin, cholest-5-en-3 beta-ol, cholesteryl alcohol, cholestrin, cordulan, dusoline, dusoran, provitamin d, cholesterine
PubChem CID 5997
ChEBI CHEBI:16113
IUPAC Name (3 S,8 S,9 S,10 R,13 R,14 S,17 R)-10,13-dimethyl-17-[(2 R)-6-methylheptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1 H-cyclopenta[a]phenanthren-3-ol
SMILES CC(C)CCCC(C)C1CCC2C1(CCC3C2CC=C4C3(CCC(C4)O)C)C

Specifications

Melting Point 147°C to 150°C
Density 1.067
Boiling Point 360°C
Packaging Plastic bottle
Quantity 100 g
Beilstein 1915888
Specific Gravity 1.067
Formula Weight 386.65
Physical Form Powder
Chemical Name or Material Cholesterol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.