accessibility menu, dialog, popup

UserName

CAS RN 57-88-5

H3CCH3(R)CH3(R)(S)(S)(S)OH(R)H3C(S)(R)H3C

CAS RN 57-88-5

IUPAC Name: (1R,3aS,3bS,7S,9aR,9bS,11aR)-9a,11a-dimethyl-1-[(2R)-6-methylheptan-2-yl]-1H,2H,3H,3aH,3bH,4H,6H,7H,8H,9H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-ol
Synonyms: cholesterol liquid crystal CN/9 20-iso-cholesterol cholesterol preparation cordulan lidinit 5:6-cholesten-3β-ol (20bFh)-cholest-5-en-3b-ol cholest-5-en-3-ol dythol
Molecular Weight (g/mol): 386.66
Molecular Formula: C27H46O
InChi Key: HVYWMOMLDIMFJA-DPAQBDIFSA-N
SMILES: CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (4)
  • (1)
Percent Purity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (6)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (4)
  • (1)
  • (4)
  • (2)
  • (1)
  • (1)