missing translation for 'onlineSavingsMsg'
Learn More

Choline chloride, Dr. Ehrenstorfer

Product Code. 30319525
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30319525 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30319525 Supplier Dr. Ehrenstorfer Supplier No. DREC11665450
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Choline Chloride
CAS 67-48-1
Analyte or Component Names Choline Chloride
Shipping Condition +20°C
Molecular Formula C5 H14 N O . Cl
Certifications/Compliance ISO/IEC 17025
Synonym Choline, chloride (8CI), Ethanaminium, 2-hydroxy-N,N,N-trimethyl-, chloride (9CI), (2-Hydroxyethyl)trimethylammonium chloride, (β-Hydroxyethyl)trimethylammonium chloride, Actiron CC 6, Bilineurin chloride, Biocolina, Biocoline, COL 24, Choline chloride, Cholinium chloride, Claytreat, Hepacholine, Hormocline, Lipotril, Luridin chloride, Neocolina, Norcol 25, Paresan, ReaShure Choline, Trimethyl(2-hydroxyethyl)ammonium chloride
InChI Formula InChI=1S/C5H14NO.ClH/c1-6(2,3)4-5-7;/h7H,4-5H2,1-3H3;1H/q+1;/p-1
SMILES [Cl-].C[N+](C)(C)CCO
IUPAC Name 2-hydroxyethyl(trimethyl)azanium;chloride
Molecular Weight (g/mol) 139.62
Quantity 250 mg
Formula Weight 139.0764 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.