missing translation for 'onlineSavingsMsg'
Learn More

Chromane-2-carboxylic acid, 97+%, Thermo Scientific™

Product Code. 10555333
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10555333 250 mg Amber Glass Bottle 250mg
10585493 1 g Amber Glass Bottle 1g
10682865 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10555333 Supplier Thermo Scientific Maybridge Supplier No. RJC01246CB
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 51939-71-0
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.19
MDL Number MFCD00044719
InChI Key SFLFCQJQOIZMHF-UHFFFAOYNA-N
Synonym chromane-2-carboxylic acid, chroman-2-carboxylic acid, 3,4-dihydro-2h-1-benzopyran-2-carboxylic acid, 2h-1-benzopyran-2-carboxylic acid, 3,4-dihydro, 2,3-dihydro-benzpyran-2-carboxylic acid, maybridge4_000177, 2-carboxychroman, chroman carboxylic acid, pubchem14568, 2-chroman-carboxylic acid
PubChem CID 2723665
IUPAC Name 3,4-dihydro-2H-chromene-2-carboxylic acid
SMILES OC(=O)C1CCC2=CC=CC=C2O1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 178.19
Percent Purity 97%
Chemical Name or Material Chromane-2-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.