missing translation for 'onlineSavingsMsg'
Learn More

Chromium Isotopic Standard: Cr @ 10 μg/mL in 2% HNO3 (Enriched in Cr-50), VHG™

Product Code. 17964356
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17964356 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17964356 Supplier VHG Labs Supplier No. VHGLIS50CR50
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Chromium Isotopic Standard: Cr @ 10 μg/mL in 2% HNO3 (Enriched in Cr-50)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Chromium 50
CAS 14304-94-0
Methods ICP-MS
Analyte or Component Names Chromium 50
Shipping Condition Room Temperature
Molecular Formula 50Cr
Synonym 50Cr
SMILES [50Cr]
IUPAC Name Chromium 50
Solvent or Matrix Dilute HNO3
Molecular Weight (g/mol) 49.95
Quantity 50 mL
Formula Weight 49.946 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.