missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chromogranin B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
216.00 € - 326.00 €
Specifications
| Antigen | Chromogranin B |
|---|---|
| Dilution | Immunocytochemistry/ Immunofluorescence 1:100-1:500 |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18680672
|
Novus Biologicals
NBP2-97350-100ul |
100 μg |
326.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18661635
|
Novus Biologicals
NBP2-97350-50ul |
50 μg |
216.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chromogranin B Polyclonal antibody specifically detects Chromogranin B in Human samples. It is validated for Immunocytochemistry/ ImmunofluorescenceSpecifications
| Chromogranin B | |
| Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cell Biology, Cytokines and Growth Factors, Neuroscience, Signal Transduction | |
| PBS (pH 7.0) | |
| 1114 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Immunocytochemistry/ Immunofluorescence 1:100-1:500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| CgB, chromogranin B (secretogranin 1), Chromogranin-B, SCG1sgI, secretogranin B, Secretogranin I, secretogranin-1, SgI | |
| Produced in rabbits immunized with E. coli-derived Human Chromogranin B fragment. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title