missing translation for 'onlineSavingsMsg'
Learn More

Chrysin, MedChemExpress

Product Code. 16481419
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16481419 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16481419 Supplier Medchem Express Supplier No. HY14589
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Chrysin is one of the most well known estrogen blockers.

Chrysin is one of the most well known estrogen blockers.

Specifications

Chemical Name or Material Chrysin
CAS 480-40-0
Color Yellow
Purity Grade Notes Research
Molecular Formula C15H10O4
Quantity 1 g
Synonym 5,7-Dihydroxyflavone
Solubility Information DMSO : ≥ 100 mg/mL (393.33 mM)
SMILES OC1=C2C(OC(C3=CC=CC=C3)=CC2=O)=CC(O)=C1
Molecular Weight (g/mol) 254.24
Formula Weight 254.24
Percent Purity 99.55%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.