missing translation for 'onlineSavingsMsg'
Learn More

Chrysin, TRC

Product Code. 30003698
Change view
Click to view available options
Quantity:
100 mg
5 g
25 g
Unit Size:
100mg
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30003698 5 g 5g
30138176 25 g 25g
30134917 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30003698 Supplier Toronto Research Chemicals Supplier No. TRCC4327305G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 480-40-0
Formula Weight 254.0579
InChI Formula InChI=1S/C15H10O4/c16-10-6-11(17)15-12(18)8-13(19-14(15)7-10)9-4-2-1-3-5-9/h1-8,16-17H
IUPAC Name 5,7-dihydroxy-2-phenylchromen-4-one
Molecular Formula C15 H10 O4
Molecular Weight (g/mol) 254.24
SMILES Oc1cc(O)c2C(=O)C=C(Oc2c1)c3ccccc3
Synonym 5,7-Dihydroxyflavone, Chrysin (6CI), Flavone, 5,7-dihydroxy- (7CI,8CI), 5,7-Dihydroxy-2-phenyl-4H-1-benzopyran-4-one, 5,7-Dihydroxy-2-phenyl-4H-chromen-4-one, 5,7-Dihydroxy-2-phenylchromen-4-one, 5,7-Dihydroxyflavone, Crysin, NSC 407436, 4H-1-Benzopyran-4-one, 5,7-dihydroxy-2-phenyl-
Chemical Name or Material Chrysin

Specifications

Quantity 5 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.