accessibility menu, dialog, popup

UserName

HOOHOO

CAS RN 480-40-0

IUPAC Name: 5,7-dihydroxy-2-phenyl-4H-chromen-4-one
Synonyms: chrysin 5,7-hydroxyflavone chrysine
Molecular Weight (g/mol): 254.24
Molecular Formula: C15H10O4
InChi Key: RTIXKCRFFJGDFG-UHFFFAOYSA-N
SMILES: OC1=CC(O)=C2C(=O)C=C(OC2=C1)C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (8)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (3)
  • (4)
Percent Purity 
  • (2)
  • (3)
  • (1)
Physical Form 
  • (3)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (1)
  • (8)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (4)
  • (2)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)