missing translation for 'onlineSavingsMsg'
Learn More

Chrysophanol, MedChemExpress

Product Code. 16496236
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16496236 100 mg 100mg
16421927 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16496236 Supplier Medchem Express Supplier No. HY13595

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Chrysophanol (Chrysophanic acid) is a natural anthraquinone, which inhibits EGF-induced phosphorylation of EGFR and suppresses activation of AKT and mTOR/p70S6K.

Chrysophanol (Chrysophanic acid) is a natural anthraquinone, which inhibits EGF-induced phosphorylation of EGFR and suppresses activation of AKT and mTOR/p70S6K.

Specifications

Chemical Name or Material Chrysophanol
CAS 481-74-3
Color Orange
Purity Grade Notes Research
Molecular Formula C15H10O4
Quantity 100 mg
Synonym Chrysophanic acid
Solubility Information DMSO : 2 mg/mL (7.87 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C1C2=C(C=CC=C2O)C(C3=CC(C)=CC(O)=C13)=O
Molecular Weight (g/mol) 254.24
Formula Weight 254.24
Percent Purity 98.07%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.