missing translation for 'onlineSavingsMsg'
Learn More

Chrysophenine (Technical Grade), TRC

Produktkod. 30366424
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mg
500 mg
2.5 g
Förpackningsstorlek:
2.5g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Quantity Förpackningsstorlek
30366424 2.5 g 2.5g
30357962 250 mg -
30351186 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 missing translation for 'options'
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30366424 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCC4331232.5G
 Restricted Item
Estimated Shipment: 22-10-2026
Lägg i varukorgen
Lägg i varukorgen

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 2870-32-8
Molecular Formula C30 H26 N4 O8 S2 . 2 Na
Molecular Weight (g/mol) 680.66
Synonym Benzenesulfonic acid, 2,2'-(1,2-ethenediyl)bis[5-[(4-ethoxyphenyl)azo]-, disodium salt (9CI), C.I. Direct Yellow 12 (6CI,7CI,8CI), Airedale Yellow CHD, Amanil Chrysophenine G, Atlantic Chrysophenine YA, Aurophenine O, Benzanil Yellow CH, Benzo Yellow LG, C.I. 24895, Calcomine Brilliant Yellow, Chrome Leather Yellow CH, Chrysophenine, Chrysophenine ESP, Chrysophenine Extra, Chrysophenine G, Chrysophenine G Extra, Chrysophenine GP, Chrysophenine J, Chrysophenine KG Conc, Chrysophenine NS, Chrysophenine Y, Chrysophenine YA-CF, Chrysophenine Yellow, Chrysophrenin G, Cotton Yellow CH, Diazamine Yellow C, Diphenyl Chrysoine 3G, Diphenyl Chrysoine 3GP, Direct Chrysophenine G, Direct Frosted Yellow G, Direct Frozen Yellow G, Direct Yellow 12, Direct Yellow C, Direct Yellow CHD, Direct Yellow CV, Direct Yellow G, Erie Yellow Y, Fenamin Yellow CP, Fixanol Yellow 3G, Kayafect Yellow S, Kayaku Chrysophenine, Kayaku Chrysophenine GE, Kayaku Chrysophenine GN, Kayaku Chrysophenine GX, Mitsui Chrysophenine G, Mitsui Chrysophenine G 400, Mitsui Chrysophenine KG, NSC 47745, Nylomine Acid Yellow C 4R, Peeramine Chrysophenine YA, Pontamine Yellow CH, Shikiso Chrysophenine GX, Tertrodirect Yellow CG, Vondacel Yellow CG
IUPAC Name disodium;5-[(E)-(4-ethoxyphenyl)diazenyl]-2-[(E)-2-[4-[(E)-(4-ethoxyphenyl)diazenyl]-2-sulfonatophenyl]ethenyl]benzenesulfonate
SMILES [Na+].[Na+].CCOc1ccc(cc1)N=Nc2ccc(\C=C\c3ccc(cc3S(=O)(=O)[O-])N=Nc4ccc(OCC)cc4)c(c2)S(=O)(=O)[O-]

Specifikationer

Quantity 2.5 g
InChI Formula InChI=1S/C30H28N4O8S2.2Na/c1-3-41-27-15-11-23(12-16-27)31-33-25-9-7-21(29(19-25)43(35,36)37)5-6-22-8-10-26(20-30(22)44(38,39)40)34-32-24-13-17-28(18-14-24)42-4-2;;/h5-20H,3-4H2,1-2H3,(H,35,36,37)(H,38,39,40);;/q;2*+1/p-2/b6-5+,33-31?,34-32?;;
Formula Weight 680.0987
Chemical Name or Material Direct Yellow 12
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.