missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CIAPIN1 Monoclonal Antibody (OTI5A6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808812
This item is not returnable.
View return policy
Description
CIAPIN1 is a cytokine-induced inhibitor of apoptosis with no relation to apoptosis regulatory molecules of the BCL2 or CASP families. Expression of CIAPIN1 is dependent on growth factor stimulation.Specifications
| CIAPIN1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q6FI81 | |
| CIAPIN1 | |
| Human recombit protein fragment corresponding to amino acids 97-312 of human CIAPIN1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI5A6 | |
| Unconjugated | |
| CIAPIN1 | |
| Anamorsin, DRE2, PRO0915 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 57019 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction