missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CIAPIN1 Monoclonal Antibody (OTI5A6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808812S
This item is not returnable.
View return policy
Description
CIAPIN1 Monoclonal antibody specifically detects CIAPIN1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| CIAPIN1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9UHH9 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 51447 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI5A6 | |
| Unconjugated | |
| Ip6k2 | |
| 2810413N20Rik; AA617265; Anamorsin; anamorsin {ECO:0000255; AU021794; basic helix-loop-helix domain containing, class B, 2; basic helix-loop-helix domain containing, class B2; basic helix-loop-helix family member e40; basic helix-loop-helix family, member e40; BHLHB2; Bhlhe40; C130042M06Rik; CIAPIN1; Class B basic helix-loop-helix protein 2; class E basic helix-loop-helix protein 40; Clast5; CR8; CUA001; cytokine induced apoptosis inhibitor 1; cytokine response gene 8; cytokine-induced apoptosis inhibitor 1; cytokine-induced apoptosis inhibitor 1 {ECO:0000255; DEC1; differentially expressed in chondrocytes 1; differentially expressed in chondrocytes protein 1; differentiated embryo chondrocyte expressed gene 1; DRE2; E47 interaction protein 1; EIP1; eip1 (E47 interaction protein 1); enhancer-of-split and hairy-related protein 2; Fe-S cluster assembly protein DRE2 homolog; fe-S cluster assembly protein DRE2 homolog {ECO:0000255; HAMAP-Rule:MF_03115}; HLHB2; predicted protein of HQ0915; | |
| Human recombinant protein fragment corresponding to amino acids 1-250 of human IP6K2 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction