missing translation for 'onlineSavingsMsg'
Learn More

Ciclopirox olamine, MedChemExpress

Product Code. 16458316
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16458316 100 mg 100mg
16444367 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16458316 Supplier Medchem Express Supplier No. HYB0450A
 Restricted Item
Estimated Shipment: 05-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Ciclopirox olamine (Ciclopirox ethanolamine) is a synthetic antifungal agent that can be used for superficial mycoses reseaech. Ciclopirox olamine has a very broad spectrum of activity and inhibits dermatophytes, yeasts, molds, and many Gram-positive and Gram-negative species pathogenic.

Ciclopirox olamine (Ciclopirox ethanolamine) is a synthetic antifungal agent that can be used for superficial mycoses reseaech. Ciclopirox olamine has a very broad spectrum of activity and inhibits dermatophytes, yeasts, molds, and many Gram-positive and Gram-negative species pathogenic.

Specifications

Chemical Name or Material Ciclopirox olamine
Physical Form Solid
CAS 41621-49-2
Color Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C14H24N2O3
For Use With (Application) COVID-19-immunoregulation
Synonym Ciclopirox ethanolamine HOE 296
Solubility Information DMSO : 25 mg/mL (93.16 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C1C=C(C)C=C(C2CCCCC2)N1O.NCCO
Molecular Weight (g/mol) 268.35
Formula Weight 268.35
Percent Purity 98.41%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.