missing translation for 'onlineSavingsMsg'
Learn More

CID-1067700, MedChemExpress

Product Code. 16458387
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
This item is not returnable. View return policy

Product Code. 16458387

Brand: Medchem Express HY13452

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress CID-1067700 (ML282) is a pan GTPase inhibitor, and competitively inhibits Ras-related in brain 7 (Rab7) with a Ki of 13 nM.

CID-1067700 (ML282) is a pan GTPase inhibitor, and competitively inhibits Ras-related in brain 7 (Rab7) with a Ki of 13 nM.

Specifications

Chemical Name or Material CID-1067700
Physical Form Powder
CAS 314042-01-8
Color Yellow
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C18H18N2O4S2
For Use With (Application) COVID-19-immunoregulation
Synonym ML282
Solubility Information DMSO : 75 mg/mL (192.07 mM; Need ultrasonic)
SMILES O=C(C1=C(NC(NC(C2=CC=CC=C2)=O)=S)SC3=C1CC(C)(C)OC3)O
Molecular Weight (g/mol) 390.48
Formula Weight 390.48
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.