missing translation for 'onlineSavingsMsg'
Learn More

Cilomilast, MedChemExpress

Product Code. 16470507
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16470507 50 mg 50mg
16464158 10 mg 10mg
16436867 5 mg 5mg
16435126 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16470507 Supplier Medchem Express Supplier No. HY10790
 Restricted Item
Estimated Shipment: 03-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Cilomilast (SB-207499) is a potent, selective and orally active inhibitor of Phosphodiesterase 4 (PDE4), with IC50s of ∼100 and 120 nM for LPDE4 and HPDE4, respectively.

Cilomilast shows selectivity for PDE4 over PDE1, PDE2, PDE3 and PDE5 (IC50=74, 65, >100, and 83 μM, respectively). Cilomilast has anti-inflammatory and immunomodulatory effects and can be used for thr research of asthma and chronic obstructive pulmonary disease (COPD).

Specifications

Chemical Name or Material Cilomilast
CAS 153259-65-5
Color White
Purity Grade Notes Research
Molecular Formula C20H25NO4
Quantity 50 mg
Synonym SB-207499
Solubility Information DMSO : ≥ 100 mg/mL (291.19 mM)
SMILES O=C([C@H]1CC[C@@](C#N)(CC1)C2=CC=C(OC)C(OC3CCCC3)=C2)O
Molecular Weight (g/mol) 343.42
Formula Weight 343.42
Percent Purity 99.89%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.