missing translation for 'onlineSavingsMsg'
Learn More

Cilostazol, MedChemExpress

Product Code. p-200101509
Encompass_Preferred
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
This item is not returnable. View return policy

Product Code. 16463377

Brand: Medchem Express HY17464

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Cilostazol (OPC 13013) is a potent and selective inhibitor of phosphodiesterase (PDE) 3A, the isoform of PDE 3 in the cardiovascular system, with an IC50 of 0.2 μM.

Cilostazol (OPC 13013) is a potent and selective inhibitor of phosphodiesterase (PDE) 3A, the isoform of PDE 3 in the cardiovascular system, with an IC50 of 0.2 μM.

Specifications

Chemical Name or Material Cilostazol
CAS 73963-72-1
Color White
Purity Grade Notes Research
Molecular Formula C20H27N5O2
Quantity 50 mg
Synonym OPC 13013
Solubility Information DMSO : 50 mg/mL (135.33 mM; Need ultrasonic)
SMILES O=C1NC2=C(C=C(OCCCCC3=NN=NN3C4CCCCC4)C=C2)CC1
Molecular Weight (g/mol) 369.46
Formula Weight 369.46
Percent Purity 99.83%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.