Learn More
Cimetidine, 98+%, Thermo Scientific™
H2 histamine receptor antagonist
Brand:  Thermo Scientific Alfa Aesar J62825.09
 View more versions of this product
Product Code. 15465129
  • ONLINE EXCLUSIVE PRICE
    134.00 €
    131.00 € / 10g
    Save 3.00 €
    2% Off
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Solubility
Soluble to 50 mM in water and to 100 mM in DMSO, solubility in HCl (almost transparency).
Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions
Videos
SDS Documents