Learn More
Cimetidine, 98+%, Thermo Scientific™
H2 histamine receptor antagonist
83.30 € - 266.99 €
Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15455129
SDS View Documents
Thermo Scientific Alfa Aesar
J62825.06
ONLINE EXCLUSIVE PRICE
85.00 €
83.30 € / 5g
Save 1.70 €
2% Off
View alternative products
Save Up to 
»
15465129
SDS View Documents
Thermo Scientific Alfa Aesar
J62825.09
ONLINE EXCLUSIVE PRICE
134.00 €
131.00 € / 10g
Save 3.00 €
2% Off
View alternative products
Save Up to 
»
15475129
SDS View Documents
Thermo Scientific Alfa Aesar
J62825.14
25g
ONLINE EXCLUSIVE PRICE
272.00 €
266.99 € / 25g
Save 5.01 €
1% Off
View alternative products
Save Up to 
»
Description
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Solubility
Soluble to 50 mM in water and to 100 mM in DMSO, solubility in HCl (almost transparency).
Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.