missing translation for 'onlineSavingsMsg'
Learn More

Cinanserin, TRC

Product Code. 30350460
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30350460 100 mg -
30356962 25 mg 25mg
30353993 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30350460 Supplier Toronto Research Chemicals Supplier No. TRCC441903100MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1166-34-3
Molecular Formula C20H24N2OS
Molecular Weight (g/mol) 340.48
Synonym N-[2-[[3-(Dimethylamino)propyl]thio]phenyl]-3-phenyl-2-propenamide, 2'-[[3-(Dimethylamino)propyl]thio]cinnamanilide, Cinanserin, Cinanserine, 2'-[[3-(Dimethylamino)propyl]thio]-cinnamanilide
IUPAC Name (E)-N-[2-[3-(dimethylamino)propylsulfanyl]phenyl]-3-phenylprop-2-enamide
SMILES CN(C)CCCSc1ccccc1NC(=O)\C=C\c2ccccc2

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C20H24N2OS/c1-22(2)15-8-16-24-19-12-7-6-11-18(19)21-20(23)14-13-17-9-4-3-5-10-17/h3-7,9-14H,8,15-16H2,1-2H3,(H,21,23)/b14-13+
Formula Weight 340.1609
Chemical Name or Material Cinanserin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.