missing translation for 'onlineSavingsMsg'
Learn More

Cinnamaldehyde, Extra Pure, SLR, Fisher Chemical

Product Code. 10548610 Shop All Fisher Chemical Products
Change view
Click to view available options
Packaging:
Amber glass bottle
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Packaging unitSize
10548610 Amber glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10548610 Supplier Fisher Chemical Supplier No. C/6098/08
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 104-55-2
Color Yellow
Packaging Amber glass bottle
MDL Number 7000
Physical Form Viscous Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.