missing translation for 'onlineSavingsMsg'
Learn More

Cinnamamide, 97%, predominantly trans

Product Code. 10615722 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10615722 5 g Glass bottle 5g
10158003 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10615722 Supplier Thermo Scientific Acros Supplier No. 110360050
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 621-79-4 | C9H9NO | 147.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 621-79-4
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.18
MDL Number MFCD00008033
InChI Key APEJMQOBVMLION-VOTSOKGWSA-N
Synonym cinnamamide, trans-cinnamamide, 3-phenylacrylamide, cinnamic amide, 2-propenamide, 3-phenyl, 3-phenylpropenamide, 2-benzylideneacetamide, e-cinnamamide, ginnamic acid amide, trans-cinnamoylamine
PubChem CID 5273472
ChEBI CHEBI:76320
IUPAC Name (E)-3-phenylprop-2-enamide
SMILES C1=CC=C(C=C1)C=CC(=O)N

Specifications

Melting Point 146.0°C to 150.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula C6H5CH=CHCONH2
Quantity 5 g
Beilstein 09, 587
Formula Weight 147.18
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Cinnamamide, Predominantly trans
Show More Show Less
compliance-icons
Product Identifier
  • Cinnamamide, predominantly trans
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.