missing translation for 'onlineSavingsMsg'
Learn More

Cinnamoyl chloride, 98%, predominantly trans

Product Code. 10687632 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10687632 500 g Glass bottle 500g
10416720 5 g Glass bottle 5g
10316030 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10687632 Supplier Thermo Scientific Acros Supplier No. 110395000
 Restricted Item
Estimated Shipment: 28-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 102-92-1 | C9H7ClO | 166.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102-92-1
Molecular Formula C9H7ClO
Molecular Weight (g/mol) 166.61
MDL Number MFCD00000732
InChI Key WOGITNXCNOTRLK-VOTSOKGWSA-N
Synonym cinnamoyl chloride, cinnamic acid chloride, cinnamic chloride, trans-3-phenylacryloyl chloride, 2-propenoyl chloride, 3-phenyl, cinnamoylchloride, e-cinnamoyl chloride, trans-cinnamoyl chloride, 2e-3-phenylprop-2-enoyl chloride, 3-phenyl-2-propenoyl chloride
PubChem CID 5354261
IUPAC Name (E)-3-phenylprop-2-enoyl chloride
SMILES C1=CC=C(C=C1)C=CC(=O)Cl

Specifications

Melting Point 33.0°C to 37.0°C
Color White to Yellow
Boiling Point 256.0°C to 258.0°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula C6H5CH=CHCOCl
Quantity 500 g
Merck Index 15, 2301
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in hydrocarbons and esters,soluble in dioxane,very soluble in logroin
Formula Weight 166.61
Percent Purity 98%
Physical Form Crystalline Solid
Chemical Name or Material Cinnamoyl chloride, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.