missing translation for 'onlineSavingsMsg'
Learn More

Cinnamyl alcohol, 98%

Product Code. 11499943
Change view
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11499943 250 g 250g
11489943 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11499943 Supplier Thermo Scientific Alfa Aesar Supplier No. A13025.30
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 104-54-1 | C9H10O | 134.178 g/mol

Cinnamyl alcohol is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used in perfumery and as well as a deodorant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cinnamyl alcohol is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff. It is also used in perfumery and as well as a deodorant.

Solubility
Soluble in water 1.8 g/L (20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 104-54-1
Molecular Formula C9H10O
Molecular Weight (g/mol) 134.178
MDL Number MFCD00002921
InChI Key OOCCDEMITAIZTP-QPJJXVBHSA-N
Synonym cinnamyl alcohol, cinnamic alcohol, 3-phenyl-2-propen-1-ol, 3-phenylprop-2-en-1-ol, zimtalcohol, styryl carbinol, e-3-phenylprop-2-en-1-ol, 3-phenylallyl alcohol, trans-cinnamyl alcohol, styryl alcohol
PubChem CID 5315892
ChEBI CHEBI:33227
IUPAC Name (E)-3-phenylprop-2-en-1-ol
SMILES C1=CC=C(C=C1)C=CCO

Specifikationer

Melting Point 32°C to 35°C
Density 1.044
Boiling Point 257°C to 258°C
Flash Point 126°C (258°F)
Odor Pleasant
Linear Formula C6H5CH=CHCH2OH
Quantity 250 g
Beilstein 1903999
Merck Index 14,2302
Solubility Information Soluble in water 1.8g/L (20°C).
Formula Weight 134.18
Percent Purity 98%
Chemical Name or Material Cinnamyl alcohol
Visa mer Visa mindre
compliance-icons
Produktbeteckning
  • Cinnamyl alcohol
Signalord
  • Warning
Riskkategorier
  • Skin sensitiser Category 1
Faroangivelser
  • H317-May cause an allergic skin reaction.
Skyddsangivelse
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Kompletterande information
  • MIXTURE LIST-Contain: Cinnamyl alcohol

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.