missing translation for 'onlineSavingsMsg'
Learn More

Cinnamyl Alcohol, TRC

Product Code. 30179052
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179052 10 g 10g
30179021 50 g 50g
30179143 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179052 Supplier Toronto Research Chemicals Supplier No. TRCC44211010G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-54-1
Molecular Formula C9 H10 O
Molecular Weight (g/mol) 134.18
Synonym Cinnamyl alcohol (6CI,8CI), 3-Phenyl-2-propen-1-ol, 1-Phenyl-3-hydroxy-1-propene, 1-Phenylprop-1-en-3-ol, 3-Hydroxy-1-phenylprop-1-ene, 3-Phenyl-2-propene-1-ol, 3-Phenyl-2-propenol, 3-Phenylallyl alcohol, Cinnamic alcohol, NSC 623440, NSC 8775, Styrone, Styryl alcohol, Styryl carbinol, γ-Phenylallyl alcohol
IUPAC Name 3-phenylprop-2-en-1-ol
SMILES OCC=Cc1ccccc1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C9H10O/c10-8-4-7-9-5-2-1-3-6-9/h1-7,10H,8H2
Formula Weight 134.0732
Chemical Name or Material Cinnamyl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.