missing translation for 'onlineSavingsMsg'
Learn More

Cinnamyl bromide, predominantly trans, 95%

Product Code. 11304594
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11304594 5 g 5g
11314594 25 g 25g
11324594 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11304594 Supplier Thermo Scientific Alfa Aesar Supplier No. L01420.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 4392-24-9 | C9H9Br | 197.075 g/mol

Cinnamyl bromide, is used in the synthesis of naftifine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cinnamyl bromide, is used in the synthesis of naftifine.

Solubility
Soluble in methanol. Insoluble in water.

Notes
Light Sensitive. Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4392-24-9
Molecular Formula C9H9Br
Molecular Weight (g/mol) 197.075
MDL Number MFCD00000245
InChI Key RUROFEVDCUGKHD-QPJJXVBHSA-N
Synonym cinnamyl bromide, 3-bromoprop-1-en-1-yl benzene, e-3-bromoprop-1-en-1-yl benzene, 3-bromo-1-phenyl-1-propene, 3-bromo-1-phenyl propene-1, e-3-bromoprop-1-enyl benzene, 3-bromo-propenyl-benzene, e-cinnamyl bromide, trans-cinnamyl bromide
PubChem CID 5357478
IUPAC Name [(E)-3-bromoprop-1-enyl]benzene
SMILES C1=CC=C(C=C1)C=CCBr

Specifications

Melting Point 26°C to 29°C
Boiling Point 88°C to 90°C (1 mmHg)
Flash Point 87°C (188°F)
Refractive Index 1.621
Quantity 5 g
UN Number UN3261
Beilstein 605833
Sensitivity Light sensitive
Solubility Information Soluble in methanol. Insoluble in water.
Formula Weight 197.08
Percent Purity 95%
Chemical Name or Material Cinnamyl bromide, predominantly trans
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.