missing translation for 'onlineSavingsMsg'
Learn More

Cinnamyl chloride, 95%, stab. with sodium carbonate

Product Code. 11457027
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
This item is not returnable. View return policy

Product Code. 11457027

Brand: Thermo Scientific Alfa Aesar A14408.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2687-12-9 | C9H9Cl | 152.62 g/mol

Cinnamyl chloride is used in the enantioselective total synthesis of helioporins C and E, bioactive marine diterpenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cinnamyl chloride is used in the enantioselective total synthesis of helioporins C and E, bioactive marine diterpenes.

Solubility
Soluble in water (0.2 g/L at 20°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature: 2 - 8°C
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2687-12-9

Specifications

Melting Point 7°C
Density 1.096
Boiling Point 86°C to 87°C (2 mmHg)
Flash Point 79°C (174°F)
Molecular Formula C9H9Cl
Refractive Index 1.5845
MDL Number MFCD00000986
Quantity 50 g
UN Number UN2927
Beilstein 741974
Synonym cinnamyl chloride, 3-chloroprop-1-en-1-yl benzene, 3-chloro-1-phenyl-1-propene, trans-cinnamyl chloride, 1e-3-chloroprop-1-en-1-yl benzene, benzene, 3-chloropropenyl, e-3-chloroprop-1-enyl benzene, unii-2i1adl56tx, 2i1adl56tx, cinnamylchloride
Solubility Information Soluble in water (0.2g/L at 20°C).
InChI Key IWTYTFSSTWXZFU-QPJJXVBHSA-N
SMILES C1=CC=C(C=C1)C=CCCl
IUPAC Name [(E)-3-chloroprop-1-enyl]benzene
Molecular Weight (g/mol) 152.62
PubChem CID 639658
Formula Weight 152.62
Percent Purity 95%
Chemical Name or Material Cinnamyl chloride, stabilized with sodium carbonate
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato