missing translation for 'onlineSavingsMsg'
Learn More

Cinnarizine, MP Biomedicals™

Product Code. 11453192 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11453192 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11453192 Supplier MP Biomedicals Supplier No. 0215498625
 Restricted Item
Estimated Shipment: 22-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 298-57-7
Color White
Molecular Formula C26H28N2
Quantity 25 g
Synonym cinnarizine, stugeron, dimitronal, stutgin, dimitron, folcodal, midronal, mitronal, stutgeron, glanil
InChI Key DERZBLKQOCDDDZ-JLHYYAGUSA-N
SMILES C1CN(CCN1CC=CC2=CC=CC=C2)C(C3=CC=CC=C3)C4=CC=CC=C4
IUPAC Name 1-benzhydryl-4-[(E)-3-phenylprop-2-enyl]piperazine
Molecular Weight (g/mol) 368.524
PubChem CID 1547484
ChEBI CHEBI:31403
Formula Weight 368.52
Physical Form Powder
Chemical Name or Material Cinnarizine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.