missing translation for 'onlineSavingsMsg'
Learn More

Cinoxacin, MedChemExpress

Product Code. 16478596
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16478596 100 mg 100mg
16424547 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16478596 Supplier Medchem Express Supplier No. HYB1085
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Cinoxacin (Compound 64716), a synthetic antimicrobial related to the quinolone class of orally active antibacterial agent. Cinoxacin has antibacterial activity against many gram-negative aerobic bacteria and inhibits bacterial DNA synthesis. Cinoxacin can be used for the research of urinary tract infections and bacterial prostatitis.

Cinoxacin (Compound 64716), a synthetic antimicrobial related to the quinolone class of orally active antibacterial agent. Cinoxacin has antibacterial activity against many gram-negative aerobic bacteria and inhibits bacterial DNA synthesis. Cinoxacin can be used for the research of urinary tract infections and bacterial prostatitis.

Specifications

Chemical Name or Material Cinoxacin
Physical Form Solid
CAS 28657-80-9
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C12H10N2O5
For Use With (Application) COVID-19-immunoregulation
Synonym Compound 64716
Solubility Information DMSO : 8.33 mg/mL (31.77 mM; Need ultrasonic)
SMILES O=C(C1=NN(CC)C2=C(C=C3C(OCO3)=C2)C1=O)O
Molecular Weight (g/mol) 262.22
Formula Weight 262.22
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.