missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This gene encodes a protein that is upregulated in response to various cytokines. The encoded protein may play a role in cell cycle progression. A translocation between this gene and the myeloid/lymphoid leukemia gene, resulting in expression of a chimeric protein, has been associated with acute myelomonocytic leukemia. Pseudogenes exist on chromosomes 7 and 8. Alternatively spliced transcript variants have been described.
Specifications
Specifications
| Antigen | CIP29 |
| Applications | Western Blot, Immunofluorescence, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.15 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | SARNP |
| Gene Accession No. | P82979, Q498U4, Q9D1J3 |
| Gene Alias | CIP29, HCC1, HSPC316, Nuclear protein Hcc 1, SARNP |
| Gene Symbols | Sarnp |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?