missing translation for 'onlineSavingsMsg'
Learn More

Ciprofloxacin, MedChemExpress

Product Code. 16432149
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16432149 1 g 1g
16490888 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16432149 Supplier Medchem Express Supplier No. HYB03561g
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Ciprofloxacin (Bay-09867) is a potent, orally active topoisomerase IV inhibitor. Ciprofloxacin induces mitochondrial DNA and nuclear DNA damage and lead to mitochondrial dysfunction, ROS production. Ciprofloxacin has anti-proliferative activity and induces apoptosis. Ciprofloxacin is a fluoroquinolone antibiotic, exhibiting potent antibacterial activity.

Ciprofloxacin (Bay-09867) is a potent, orally active topoisomerase IV inhibitor. Ciprofloxacin induces mitochondrial DNA and nuclear DNA damage and lead to mitochondrial dysfunction, ROS production. Ciprofloxacin has anti-proliferative activity and induces apoptosis. Ciprofloxacin is a fluoroquinolone antibiotic, exhibiting potent antibacterial activity.

Specifications

Chemical Name or Material Ciprofloxacin
Physical Form Solid
CAS 85721-33-1
Color Off-White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C17H18FN3O3
For Use With (Application) COVID-19-immunoregulation
Synonym Bay-09867
Solubility Information 0.1 M HCL : 16.67 mg/mL (50.31 mM; ultrasonic and warming and adjust pH to 2 with HCl and heat to 60°C) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C(C1=CN(C2CC2)C3=C(C=C(F)C(N4CCNCC4)=C3)C1=O)O
Molecular Weight (g/mol) 331.34
Formula Weight 331.34
Percent Purity 99.32%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.