missing translation for 'onlineSavingsMsg'
Learn More

cis-10-Hydroxy Nortriptyline, TRC

Product Code. 30004334
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004334 25 mg 25mg
30006867 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30004334 Supplier Toronto Research Chemicals Supplier No. TRCH94881525MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 47132-19-4
Molecular Formula C19 H21 N O
Molecular Weight (g/mol) 279.38
Synonym 5H-Dibenzo[a,d]cyclohepten-10-ol, 10,11-dihydro-5-[3-(methylamino)propylidene]-, (5Z)-5H-Dibenzo[a,d]cyclohepten-10-ol, 10,11-dihydro-5-[3-(methylamino)propylidene]-, (Z)-, 10-Hydroxy-(Z)-nortriptyline, cis-10-Hydroxynortriptyline, (Z)-10-Hydroxynortriptyline
IUPAC Name (11Z)-11-[3-(methylamino)propylidene]-5,6-dihydrodibenzo[1,2-a:2',1'-d][7]annulen-5-ol
SMILES CNCC\C=C/1\c2ccccc2CC(O)c3ccccc13

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C19H21NO/c1-20-12-6-11-16-15-8-3-2-7-14(15)13-19(21)18-10-5-4-9-17(16)18/h2-5,7-11,19-21H,6,12-13H2,1H3/b16-11-
Formula Weight 279.1623
Chemical Name or Material cis-10-Hydroxy Nortriptyline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.