missing translation for 'onlineSavingsMsg'
Learn More

cis-2-Pentene

Product Code. 10299573 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10299573 5 g Glass bottle 5g
10320684 1 g Glass bottle 1g
10116823 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10299573 Supplier Thermo Scientific Acros Supplier No. 383590050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 627-20-3 | C5H10 | 70.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 627-20-3
Molecular Formula C5H10
Molecular Weight (g/mol) 70.14
MDL Number MFCD00063978
InChI Key QMMOXUPEWRXHJS-HYXAFXHYSA-N
Synonym cis-2-pentene, z-2-pentene, 2-pentene, z, cis-pentene, z-pent-2-ene, unii-54ur3xz4fc, cis-pentene-2, 54ur3xz4fc, 2-pentene, 2-pentene, 2z
PubChem CID 5326160
IUPAC Name (Z)-pent-2-ene
SMILES CC\C=C/C

Specifications

Melting Point -151.0°C
Color Colorless
Density 0.6500g/mL
Boiling Point 37.0°C
Flash Point −27°C
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula CH3CHCHCH2CH3
Refractive Index 1.3810 to 1.3830 (20°C, 589nm)
Quantity 5 g
Merck Index 15, 7235
Specific Gravity 0.65
Solubility Information Solubility in water: insoluble.
Formula Weight 70.13
Percent Purity ≥95.0%
Physical Form Liquid
Chemical Name or Material cis-2-Pentene
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • cis-2-Pentene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.