missing translation for 'onlineSavingsMsg'
Learn More

cis-9,10-Dihydroxy-9,10-dihydrophenanthrene, TRC

Product Code. 30378938
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30378938 100 mg -
30374830 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30378938 Supplier Toronto Research Chemicals Supplier No. TRCD452750100MG
 Restricted Item
Estimated Shipment: 11-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2510-71-6
Molecular Formula C14H12O2
Molecular Weight (g/mol) 212.24
Synonym (9R,10S)-rel-9,10-Dihydro-9,10-dihydroxyphenanthrene, 9,10-Dihydrophenanthrene-cis-9,10-diol, Phenanthrene cis-9,10-Hihydrodiol, cis-9,10-Dihydro-9,10-phenanthrenediol
SMILES O[C@@H]1[C@H](O)c2ccccc2c3ccccc13

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C14H12O2/c15-13-11-7-3-1-5-9(11)10-6-2-4-8-12(10)14(13)16/h1-8,13-16H
Formula Weight 212.0837
Chemical Name or Material cis-9,10-Dihydroxy-9,10-dihydrophenanthrene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.