missing translation for 'onlineSavingsMsg'
Learn More

cis-Dichlorobis(triphenylphosphine)platinum(II), Premion™, 99.95% (metals basis), Pt 24.2% min

Product Code. 11349407
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11349407

Brand: Thermo Scientific Alfa Aesar 041008.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 15604-36-1 | C36H30Cl2P2Pt | 790.57 g/mol

cis-Dichlorobis(triphenylphosphine)platinum(II) is used in hydrosilylation, wax oil catalytic cracking, platinum plating and as platinum catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15604-36-1
Molecular Formula C36H30Cl2P2Pt
Molecular Weight (g/mol) 790.57
MDL Number MFCD00010825
InChI Key ILIBUWCDNDIUOT-UHFFFAOYSA-L
Synonym cis-?dichlorobis triphenylphosphine ?platinum ii
PubChem CID 131667321
IUPAC Name platinum(2+);triphenylphosphane;dihydrochloride
SMILES Cl[Pt++]Cl.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 310°C (decomposition)
Quantity 5 g
Sensitivity Hygroscopic
Solubility Information Insoluble in water. Slightly soluble in chloroform,hexane and toluene.
Formula Weight 790.58
Percent Purity 99.95%
Grade Premion
Concentration or Composition (by Analyte or Components) Pt 24.2% min.
Physical Form Powder
Assay (metals basis)
Chemical Name or Material cis-Dichlorobis(triphenylphosphine)platinum(II)
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.