missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Citalopram hydrobromide

Product Code. 15732987 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15732987 5 g 5g
15722987 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15732987 Supplier Thermo Scientific Chemicals Supplier No. 462310050
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 59729-32-7 | C20H21FN2O·HBr | 405.3 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59729-32-7
Molecular Formula C20H21FN2O·HBr
Molecular Weight (g/mol) 405.3
InChI Key WIHMBLDNRMIGDW-UHFFFAOYSA-N
Synonym citalopram hydrobromide, citalopram hbr, cipramil, celexa, 1-3-dimethylamino propyl-1-4-fluorophenyl-1,3-dihydroisobenzofuran-5-carbonitrile hydrobromide, elopram, seropram, 1-3-dimethylamino propyl-1-4-fluorophenyl-1,3-dihydro-5-isobenzofurancarbonitrile hydrobromide, citalopram hydrobromide usan
PubChem CID 77995
IUPAC Name 1-[3-(dimethylamino)propyl]-1-(4-fluorophenyl)-3H-2-benzofuran-5-carbonitrile;hydrobromide
SMILES CN(C)CCCC1(C2=C(CO1)C=C(C=C2)C#N)C3=CC=C(C=C3)F.Br

Specifications

Melting Point 185.0°C to 188.0°C
Quantity 5 g
Solubility Information (1% in ethanol) Clear colorless
Formula Weight 405.3
Chemical Name or Material Citalopram hydrobromide
compliance-icons
Product Identifier
  • Citalopram hydrobromide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: Citalopram hydrobromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.