missing translation for 'onlineSavingsMsg'
Learn More

Citalopram N-Oxide, TRC

Product Code. 30011732
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011732 5 mg 5mg
30009213 1 mg 1mg
30004256 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011732 Supplier Toronto Research Chemicals Supplier No. TRCC5050255MG
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 63284-72-0
Molecular Formula C20 H21 F N2 O2
Molecular Weight (g/mol) 340.39
Synonym 5-Isobenzofurancarbonitrile, 1-[3-(dimethyloxidoamino)propyl]-1-(4-fluorophenyl)-1,3-dihydro-, 5-Isobenzofurancarbonitrile, 1-[3-(dimethylamino)propyl]-1-(4-fluorophenyl)-1,3-dihydro-, N1-oxide, 1-[3-(Dimethyloxidoamino)propyl]-1-(4-fluorophenyl)-1,3-dihydro-5-isobenzofurancarbonitrile, Citalopram N-oxide, Lu 11-305
IUPAC Name 3-[5-cyano-1-(4-fluorophenyl)-3H-2-benzofuran-1-yl]-N,N-dimethylpropan-1-amine oxide
SMILES C[N+](C)([O-])CCCC1(OCc2cc(ccc12)C#N)c3ccc(F)cc3

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C20H21FN2O2/c1-23(2,24)11-3-10-20(17-5-7-18(21)8-6-17)19-9-4-15(13-22)12-16(19)14-25-20/h4-9,12H,3,10-11,14H2,1-2H3
Formula Weight 340.1587
Percent Purity >95%
Chemical Name or Material Citalopram N-Oxide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.