missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CKAP4 Recombinant Rabbit Monoclonal Antibody (HL2562)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA556651
This item is not returnable.
View return policy
Description
CKAP4 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)
This gene encodes the coiled-coil containing protein optineurin. Optineurin may play a role in normal-tension glaucoma and adult-onset primary open angle glaucoma. Optineurin interacts with adenovirus E3-14. 7K protein and may utilize tumor necrosis factor-alpha or Fas-ligand pathways to mediate apoptosis, inflammation or vasoconstriction. Optineurin may also function in cellular morphogenesis and membrane trafficking, vesicle trafficking, and transcription activation through its interactions with the RAB8, huntingtin, and transcription factor IIIA proteins. Alternative splicing results in multiple transcript variants encoding the same protein.
Specifications
| CKAP4 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q07065 | |
| CKAP4 | |
| Recombinant protein encompassing a sequence within the Extracellular domain of human CKAP4. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| HL2562 | |
| Unconjugated | |
| CKAP4 | |
| 5630400A09Rik; 63 kDa membrane protein; 63-kDa cytoskeleton-linking membrane protein; CKAP4; Climp-63; cytoskeleton associated protein 4; cytoskeleton-associated protein 4; ERGIC-63; LPC-1; p63; transmembrane protein (63kD), endoplasmic reticulum/Golgi intermediate compartment; type-II transmembrane protein p63 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 10970 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction