missing translation for 'onlineSavingsMsg'
Learn More

Analytes C Interference Check Standard For ICP/MS, SPEX CertiPrep™

Product Code. 15678255 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
125 mL
Unit Size:
125mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15678255 125 mL 125mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15678255 Supplier SPEX Certiprep Supplier No. CLINTB3N
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Ag2μg/mL
As2μg/mL
Ba2μg/mL
Be2μg/mL
Cd2μg/mL
Co2μg/mL
Cr2μg/mL
Cu2μg/mL
Mn2μg/mL
Ni2μg/mL
Pb2μg/mL
Sb2μg/mL
Se2μg/mL
Tl2μg/mL
V2μg/mL
Zn2μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Chemical Name or Material Analytes B3 without Mercury
Name Note For ILM05.2, ICP/MS; Without mercury
Packaging Triple-leached LDPE bottle
CAS 7440-41-7, 7732-18-5, 7440-28-0, 7440-22-4, 7440-43-9
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
CAS Max % 0.9, 0.0, 0.0, 0.0, 0.0, 0.0
Analyte or Component Names Silver,Arsenic,Barium
Technique ICP-MS
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Solution Type Standard Solution
Type ICP/ICP-MS Standard
Element Ag, As, Ba
Solvent or Matrix 2% HNO3/tr Tart. Acid/tr HF
Quantity 125 mL
Grade PPT
Concentration or Composition (by Analyte or Components) 2mg/L
Show More Show Less
compliance-icons
Product Identifier
  • Analytes C (For ILM 05.2)
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
  • MIXTURE LIST-Contain: nitric acid, hydrofluoric acid

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.