Learn More
Clarithromycin, 960-1040 ^mg/mg
Descrizione
A macrolide antibiotic that inhibits bacterial growth by targeting the 50S ribosomal subunit
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- A macrolide antibiotic that inhibits bacterial growth by targeting the 50S ribosomal subunit
Specifica
Specifica
| Type | Clarithromycin |
| Physical Form | Powder |
| Melting Point | 217°C to 220°C |
| Color | White |
| Quantity | 100 mg |
| Flash Point | >110°C (230°F) |
| Boiling Point | 86°C |
| Merck Index | 14,2339 |
| Solubility Information | Soluble in DMSO |
| Formula Weight | 747.95 |
| Vedi altri risultati |
RUO – Research Use Only
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.