missing translation for 'onlineSavingsMsg'
Learn More

Clarithromycin, MedChemExpress

Product Code. 16491266
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16491266 200 mg 200mg
16477346 100 mg 100mg
16431876 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16491266 Supplier Medchem Express Supplier No. HY17508
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Clarithromycin has a broad spectrum of antimicrobial activity. Clarithromycin inhibits the CYP3A4-catalyzed triazolam alpha-hydroxylation with the IC50 (Ki) value of 56 (43) μM. Clarithromycin significantly inhibits the HERG potassium current.Clarithromycin affects the autophagic flux by impairing the signaling pathway linking hERG1 and PI3K.

Clarithromycin has a broad spectrum of antimicrobial activity. Clarithromycin inhibits the CYP3A4-catalyzed triazolam alpha-hydroxylation with the IC50 (Ki) value of 56 (43) μM. Clarithromycin significantly inhibits the HERG potassium current.Clarithromycin affects the autophagic flux by impairing the signaling pathway linking hERG1 and PI3K.

Specifications

Chemical Name or Material Clarithromycin
Physical Form Solid
CAS 81103-11-9
Color White
Quantity 200 mg
Purity Grade Notes Research
Molecular Formula C38H69NO13
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 33.33 mg/mL (44.56 mM; Need ultrasonic)
SMILES C[C@@](OC)(C[C@H](C([C@@H]1C)=O)C)[C@@H]([C@H]([C@@H]([C@H](C(O[C@H](CC)[C@@](C)(O)[C@@H]1O)=O)C)O[C@@](O[C@@H](C)[C@@H]2O)([H])C[C@@]2(C)OC)C)O[C@@](O[C@H](C)C[C@@H]3N(C)C)([H])[C@@H]3O
Molecular Weight (g/mol) 747.95
Formula Weight 747.95
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.