missing translation for 'onlineSavingsMsg'
Learn More

TOKU-E™ Clarithromycin ReadyMade™ Solution

Product Code. 30044048
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mL
10 x 1 mL
Unit Size:
10mL
1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30044048 1 mL 1mL
30044047 10 x 1 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30044048 Supplier TOKU-E™ Supplier No. C2881ML

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Clarithromycin ReadyMade™ Solution is provided as a filter-sterilized solution of Clarithromycin in water.

  • CAS Number: 81103-11-9
  • Molecular Formula: C38H69NO13
  • Molecular Weight: 747.95 g/mol
  • Storage Conditions: -20°C
  • Form: Liquid
  • Source: Semi-synthetic
  • Spectrum: Clarithromycin is a broad-spectrum antibiotic with bacteriostatic action on a wide range of Gram-positive and Gram-negative bacteria including anaerobes. It is also effective for Mycoplasma and Mycobacteria
  • Mechanism of Action: Macrolide antibiotics inhibit bacterial growth by targeting the 50S ribosomal subunit preventing peptide bond formation and translocation during protein synthesis

Specifications

Chemical Name or Material Clarithromycin
Physical Form Liquid
CAS 81103-11-9
Source Semi-synthetic
Packaging 2 mL plastic vial
Sterile Filtered Yes, 0.22 μm
Sterility Sterile
Color Clear Colorless to Pale Yellow
Concentration 1 mg/mL
Quantity 1 mL
Purity Grade Notes >95% (HPLC)
Assay Percent Range 96.0 to 102%
Assay Percent Notes Clarithromycin, Dried Basis
Molecular Formula C38H69NO13
Product Line ReadyMade™ Solutions
Shipping Condition Ice Pack
Synonym 6-O-Methylerythromycin; 6-O-Methylerythromycin A; Clathromycin
Solubility Information Soluble in Water
IUPAC Name (3R,4S,5S,6R,7R,9R,11R,12R,13S,14R)-6-[(2S,3R,4S,6R)-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy-14-ethyl-12,13-dihydroxy-4-[(2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyloxan-2-yl]oxy-7-methoxy-3,5,7,9,11,13-hexamethyl-oxacyclotetradecane-2,10-dione
Molecular Weight (g/mol) 747.95 g/mol
Formula Weight 747.95 amu
Percent Purity >95%
Grade Antibiotic
Concentration or Composition (by Analyte or Components) Clarithromycin 0.1%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.